Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Aminoheptane, ChiPros 99+%, ee 98+%

Chemical Identifiers

Chemical Name or Material (R)-(-)-2-Aminoheptane
CAS 6240-90-0
Molecular Formula C7H17N
Formula Weight 115.22
MDL Number MFCD00066323
UN Number UN2733
Beilstein 4652352
Synonym (2R)-(-)-Heptylamine; (R)-1-Methylhexylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1930603

 
alfa aesar™
L1930603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930606

 
alfa aesar™
L1930606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 115.22
Chemical Name or Material (R)-(-)-2-Aminoheptane
Assay ≥99%
Molecular Formula C7H17N
MDL Number MFCD00066323
CAS 6240-90-0
UN Number UN2733
Synonym (2R)-(-)-Heptylamine; (R)-1-Methylhexylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay Percent Notes ee ≥98%
Beilstein 4652352
pH 11.45
Boiling Point 142° to 144°C
Quantity 1g
Flash Point 54°C (129°F)
Odor Amine-like
Optical Rotation −7.2° (Neat)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.