Please login to your online account to display your discounted pricing

(R)-2-Aminobut-3-en-1-ol, benzoate salt, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-2-Aminobut-3-en-1-ol, benzoate salt
Name Note 95%, (98% e.e.)
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H15NO3
Formula Weight 209.24
MDL Number MFCD01631086
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363160010

  SDS
thermo fisher scientific
363160010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC363165000

  SDS
thermo fisher scientific
363165000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 209.24
Quantity 1g
Color White to almost white
TSCA TSCA
Chemical Name or Material (R)-2-Aminobut-3-en-1-ol, benzoate salt
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C11H15NO3
MDL Number MFCD01631086
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (GC)
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.