Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Amino-5-phosphonopentanoic acid, 99%

A competitive NMDA antagonist

Chemical Identifiers

Chemical Name or Material (R)-(-)-2-Amino-5-phosphonopentanoic acid
CAS 79055-68-8
Molecular Formula C5H12NO5P
Formula Weight 197.13
MDL Number MFCD00078839
Synonym D-AP5; D-2-Amino-5-phosphonovaleric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64887MCR

  SDS
alfa aesar™
J64887MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.13
Chemical Name or Material (R)-(-)-2-Amino-5-phosphonopentanoic acid
Molecular Formula C5H12NO5P
CAS 79055-68-8
Physical Form Powder
Assay 99%
Sensitivity Light Sensitive
MDL Number MFCD00078839
Synonym D-AP5; D-2-Amino-5-phosphonovaleric acid
Health Hazard 1 Warning
Optical Rotation −22° (c=1 in 6N HCl)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Quantity 1mg
Recommended Storage 2° to 8°C
RTECS Number RD2513000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C