missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Amino-3-methylbutane, ChiPros 98%, ee 97%

$155.76 - $1,481.97

Chemical Identifiers

CAS 34701-33-2
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD01075731
InChI Key JOZZAIIGWFLONA-RXMQYKEDSA-N
Synonym r---2-amino-3-methylbutane, r---3-methyl-2-butylamine, r-3-methyl-2-butylamine, r-3-methylbutan-2-amine, 2r-3-methylbutan-2-amine, unii-s9ygl88qkq, s9ygl88qkq, 2r-3-methyl-butan-2-amine, 2-butanamine, 3-methyl-, r, r-3-methyl-2-butanamine
PubChem CID 6999786
IUPAC Name (2R)-3-methylbutan-2-amine
SMILES CC(C)C(C)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1633003 View Documents Alfa Aesar™
L1633003
1g Each for $155.76
 
AAL1633006 View Documents Alfa Aesar™
L1633006
5g Each for $485.77
 
AAL1633014 View Documents Alfa Aesar™
L1633014
25g Each for $1,481.97
 
Description

Description

Applications
(R)-(-)-2-Amino-3-methylbutane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water (60g/L at 20°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under dry inert gas. Incompatible with oxidizing agents. Store in cool place. It is sensitive to air.
Specifications

Chemical Identifiers

34701-33-2
87.166
JOZZAIIGWFLONA-RXMQYKEDSA-N
6999786
CC(C)C(C)N
C5H13N
MFCD01075731
r---2-amino-3-methylbutane, r---3-methyl-2-butylamine, r-3-methyl-2-butylamine, r-3-methylbutan-2-amine, 2r-3-methylbutan-2-amine, unii-s9ygl88qkq, s9ygl88qkq, 2r-3-methyl-butan-2-amine, 2-butanamine, 3-methyl-, r, r-3-methyl-2-butanamine
(2R)-3-methylbutan-2-amine

Specifications

<21°C (69°F)
85°C to 87°C
34701-33-2
MFCD01075731
1718798
JOZZAIIGWFLONA-RXMQYKEDSA-N
(2R)-3-methylbutan-2-amine
6999786
98%
ee 97%
−5.2° (Neat)
1g
C5H13N
UN2733
r---2-amino-3-methylbutane, r---3-methyl-2-butylamine, r-3-methyl-2-butylamine, r-3-methylbutan-2-amine, 2r-3-methylbutan-2-amine, unii-s9ygl88qkq, s9ygl88qkq, 2r-3-methyl-butan-2-amine, 2-butanamine, 3-methyl-, r, r-3-methyl-2-butanamine
CC(C)C(C)N
87.166
87.16
(R)-(-)-2-Amino-3-methylbutane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H311-H314-H318-H302-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P280-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

RTECSNumber : UI1100000