missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-2-Amino-3-methyl-1-butanol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC422470050*

 View more versions of this product

Catalog No. AC422470050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(R)-(-)-2-Amino-3-methyl-1-butanol
4276-09-9
97.5
Glass bottle
0.9310g/mL
32° to 36°C
Low Melting Crystals or Crystalline Mass
5g
C5H13NO
MFCD00064297
98%
Authentic
100.0
-15° to -18° (20°C, 589nm) (c=10, EtOH)
−16.50 (20.00°C c=10,EtOH)
White to light yellow
≥97.5%
98%
103.16
D-Valinol

Chemical Identifiers

4276-09-9
100.0
C5H13NO
MFCD00064297
(R)-(-)-2-Amino-3-methyl-1-butanol, 98%
97.5
98%
103.16
D-Valinol
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Recommended Storage : Refrigerator +4°C