Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol hydrochloride, 98%

$502.73 - $502.73

Chemical Identifiers

Chemical Name or Material (R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol hydrochloride
CAS 56755-20-5
Molecular Formula C17H21NO·HCl
Formula Weight 291.85g/mol
MDL Number MFCD01318247
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5106906

 
alfa aesar™
H51069-06
Each for $502.73
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material (R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol hydrochloride
CAS 56755-20-5
Assay 98%
Molecular Formula C17H21NO·HCl
Formula Weight 291.85g/mol
MDL Number MFCD01318247
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.