missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-2-Amino-1-propanol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC400440050

 View more versions of this product

Catalog No. AC400440050


 

Specifications Description & Specifications

Specifications

(R)-(-)-2-Amino-1-propanol
Authentic
99% min. (GC)
Glass bottle
-16° to -18° (20°C, 589nm) (neat)
− 17.00 (20.00°C c=neat)
Oily Liquid
≥97.5%
C3H9NO
75.11
04, III, 735
35320-23-1
97.5
100.0
1.4480 to 1.4510 (20°C, 589nm)
0.9650g/mL
Clear colorless to light yellow
5g
98%
CH3CH(NH2)CH2OH
MFCD00064413
D-Alaninol

Chemical Identifiers

35320-23-1
100.0
C3H9NO
75.11
D-Alaninol
≥97.5%
97.5
98%
CH3CH(NH2)CH2OH
MFCD00064413
(R)-(-)-2-Amino-1-propanol

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger