missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-2-Amino-1-diphenylphosphino-3-methylbutane, 97+%

Chemical Identifiers

CAS 1400149-69-0
Molecular Formula C17H22NP
Molecular Weight (g/mol) 271.344
MDL Number MFCD17013992
InChI Key ZZLCXURCZWQECA-KRWDZBQOSA-N
Synonym r-1-diphenylphosphino-2-amino-3-methylbutane, r-1-diphenylphosphino-3-methyl-2-butanamine, 2r-1-diphenylphosphino-3-methyl-2-butanamine, 2r-2-amino-3-methylbutyl diphenylphosphane, r-1-diphenylphosphino-3-methyl-2-butylamine, r-1-diphenylphosphino-3-methyl-2-butylamine, r-1-diphenylphosphino-3-methylbutan-2-amine, r-2-amino-1-diphenylphosphino-3-methylbutane
PubChem CID 72376444
IUPAC Name (2R)-1-diphenylphosphanyl-3-methylbutan-2-amine
SMILES CC(C)C(CP(C1=CC=CC=C1)C2=CC=CC=C2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60004MD View Documents Alfa Aesar™
H60004MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6000403 View Documents Alfa Aesar™
H6000403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1400149-69-0
271.344
ZZLCXURCZWQECA-KRWDZBQOSA-N
72376444
CC(C)C(CP(C1=CC=CC=C1)C2=CC=CC=C2)N
C17H22NP
MFCD17013992
r-1-diphenylphosphino-2-amino-3-methylbutane, r-1-diphenylphosphino-3-methyl-2-butanamine, 2r-1-diphenylphosphino-3-methyl-2-butanamine, 2r-2-amino-3-methylbutyl diphenylphosphane, r-1-diphenylphosphino-3-methyl-2-butylamine, r-1-diphenylphosphino-3-methyl-2-butylamine, r-1-diphenylphosphino-3-methylbutan-2-amine, r-2-amino-1-diphenylphosphino-3-methylbutane
(2R)-1-diphenylphosphanyl-3-methylbutan-2-amine

Specifications

>110°C (230°F)
1400149-69-0
MFCD17013992
r-1-diphenylphosphino-2-amino-3-methylbutane, r-1-diphenylphosphino-3-methyl-2-butanamine, 2r-1-diphenylphosphino-3-methyl-2-butanamine, 2r-2-amino-3-methylbutyl diphenylphosphane, r-1-diphenylphosphino-3-methyl-2-butylamine, r-1-diphenylphosphino-3-methyl-2-butylamine, r-1-diphenylphosphino-3-methylbutan-2-amine, r-2-amino-1-diphenylphosphino-3-methylbutane
CC(C)C(CP(C1=CC=CC=C1)C2=CC=CC=C2)N
271.344
271.34
(R)-2-Amino-1-diphenylphosphino-3-methylbutane
250mg
C17H22NP
UN2735
ZZLCXURCZWQECA-KRWDZBQOSA-N
(2R)-1-diphenylphosphanyl-3-methylbutan-2-amine
72376444
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.