missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-2-Amino-1-diphenylphosphino-3,3-dimethylbutane, 97+%

Manufacturer:  Alfa Aesar™ H6050803

 View more versions of this product

Catalog No. AAH6050803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1.026
1.589
1g
C18H24NP
UN2735
SGBNMEAXYLJQRV-KRWDZBQOSA-N
(2R)-1-diphenylphosphanyl-3,3-dimethylbutan-2-amine
72376445
≥97%
>110°C (230°F)
Air Sensitive
1366384-12-4
MFCD17013994
2r-1-diphenylphosphino-3,3-dimethyl-2-butana, 2r-2-amino-3,3-dimethylbutyl diphenylphosphane, r-1-diphenylphosphino-2-amino-3,3-dimethylbutane, r-1-diphenylphosphino-3,3-dimethyl-2-butanamine, r-1-diphenylphosphino-3,3-dimethyl-2-butylamine, r-1-diphenylphosphino-3,3-dimethylbutan-2-amine, r-2-amino-1-diphenylphosphino-3,3-dimethylbutane
CC(C)(C)C(CP(C1=CC=CC=C1)C2=CC=CC=C2)N
285.371
285.36

Chemical Identifiers

1366384-12-4
285.371
SGBNMEAXYLJQRV-KRWDZBQOSA-N
72376445
CC(C)(C)C(CP(C1=CC=CC=C1)C2=CC=CC=C2)N
C18H24NP
MFCD17013994
2r-1-diphenylphosphino-3,3-dimethyl-2-butana, 2r-2-amino-3,3-dimethylbutyl diphenylphosphane, r-1-diphenylphosphino-2-amino-3,3-dimethylbutane, r-1-diphenylphosphino-3,3-dimethyl-2-butanamine, r-1-diphenylphosphino-3,3-dimethyl-2-butylamine, r-1-diphenylphosphino-3,3-dimethylbutan-2-amine, r-2-amino-1-diphenylphosphino-3,3-dimethylbutane
(2R)-1-diphenylphosphanyl-3,3-dimethylbutan-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents