missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-2-Amino-1-butanol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC244035000

 View more versions of this product

Catalog No. AC244035000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(R)-(-)-2-Amino-1-butanol
90°C
Glass bottle
0.947
-10
White
500mL
97.5
97.5% min. (GC)
C2H5CH(NH2)CH2OH
04,III,771
1-butanol, 2-amino-, 2r, 1-butanol, 2-amino-, r, 2-amino-1-butanol, r, 2-amino-1-butanol,-, 2r-2-aminobutan-1-ol, l---2-amino-1-butanol, r---2-amino-1-butanol, r-2-amino-1-butanol, r-2-aminobutan-1-ol, unii-4f725v55ln
JCBPETKZIGVZRE-SCSAIBSYSA-N
(2R)-2-aminobutan-1-ol
2723856
Powder, Crystals and/or Chunks
0.9470g/mL
Authentic
1.4495 to 1.4535
−10° (19°C c=neat)
172°C to 174°C
-2°C
5856-63-3
100.0
C4H11NO
MFCD00064419
10, 429
Solubility in water: miscible. Other solubilities: very soluble in most of the usual solvents,hardly soluble in aliphatic hydrocarbons
CCC(CO)N
89.14
89.14
98%

Chemical Identifiers

5856-63-3
89.14
JCBPETKZIGVZRE-SCSAIBSYSA-N
2723856
CCC(CO)N
C4H11NO
MFCD00064419
1-butanol, 2-amino-, 2r, 1-butanol, 2-amino-, r, 2-amino-1-butanol, r, 2-amino-1-butanol,-, 2r-2-aminobutan-1-ol, l---2-amino-1-butanol, r---2-amino-1-butanol, r-2-amino-1-butanol, r-2-aminobutan-1-ol, unii-4f725v55ln
(2R)-2-aminobutan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 227-476-4

SDS Documents