Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2-Acetoxysuccinic anhydride, 98%

$130.03 - $492.72

Chemical Identifiers

Chemical Name or Material (R)- (+)-2-Acetoxysuccinic anhydride
CAS 79814-40-7
Molecular Formula C6H6O5
Formula Weight 158.11
MDL Number MFCD00145214
Synonym D-O-Acetylmalic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4436303

  SDS
alfa aesar™
4436303
1g Each for $130.03

AA4436306

  SDS
alfa aesar™
4436306
5g Each for $492.72
Description & Specifications

Specifications

Formula Weight 158.11
CAS 79814-40-7
Quantity 1g
Melting Point 56° to 58°C
MDL Number MFCD00145214
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (R)- (+)-2-Acetoxysuccinic anhydride
Assay 98%
Sensitivity Moisture Sensitive
Molecular Formula C6H6O5
Health Hazard 1 Warning
Physical Form Powder
Synonym D-O-Acetylmalic anhydride
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.