missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(-)-2,3-O-Isopropylideneglycerol, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2303706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(R)-(-)-2,3-O-Isopropylideneglycerol | |
1.066 | |
80°C (176°F) | |
1.435 | |
5 g | |
14,5213 | |
RNVYQYLELCKWAN-RXMQYKEDSA-N | |
CC1(OCC(O1)CO)C | |
132.159 | |
132.18 |
14347-78-5 | |
82°C to 83°C (14 mmHg) | |
C6H12O3 | |
MFCD00003213 | |
80117 | |
r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerol | |
−11.5° (c=5 in Methanol) | |
[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol | |
736056 | |
96% |
Chemical Identifiers
14347-78-5 | |
132.159 | |
RNVYQYLELCKWAN-RXMQYKEDSA-N | |
736056 | |
CC1(OCC(O1)CO)C |
C6H12O3 | |
MFCD00003213 | |
r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerol | |
[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
RTECSNumber : JI0400000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only