Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt, 95%

Chemical Identifiers

Chemical Name or Material (R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt
CAS 201472-66-4
Formula Weight 485.66
MDL Number MFCD00238312
Synonym Boc-Dab(Boc)-OH.DCHA; (R)-Nα,Nγ-Di-Boc-2,3-diaminopropionic acid dicyclohexylammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6228303

 
alfa aesar™
H6228303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6228306

 
alfa aesar™
H6228306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62283MD

 
alfa aesar™
H62283MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 485.66
CAS 201472-66-4
Quantity 1g
Chemical Name or Material (R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt
Physical Form Solid
MDL Number MFCD00238312
Synonym Boc-Dab(Boc)-OH.DCHA; (R)-Nα,Nγ-Di-Boc-2,3-diaminopropionic acid dicyclohexylammonium salt
Assay 95%