missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt, 95%

Manufacturer:  Alfa Aesar™ H62283MD

 View more versions of this product

Catalog No. AAH62283MD



Specifications

Specifications

(R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt
201472-66-4
MFCD00238312
BGTQRELDRNOEDA-FVGYRXGTSA-N
(2S)-2,4-bis[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid;N-cyclohexylcyclohexanamine
56777023
Solid
250mg
C26H49N3O6
2s-2,4-bis tert-butoxycarbonyl amino butanoic acid n,n-dicyclohexylamine salt
CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
499.693
485.66
95%

Chemical Identifiers

201472-66-4
499.693
BGTQRELDRNOEDA-FVGYRXGTSA-N
56777023
CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
C26H49N3O6
MFCD00238312
2s-2,4-bis tert-butoxycarbonyl amino butanoic acid n,n-dicyclohexylamine salt
(2S)-2,4-bis[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid;N-cyclohexylcyclohexanamine
Documents