Please login to your online account to display your discounted pricing

(R)-(-)-2,2-Dimethyl-4-(hydroxymethyl)-1,3-dioxolane-p-toluenesulfonate, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  133750010

 View more versions of this product

Catalog No. AC133750010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Color Clear light yellow to light brownor white to tan
Physical Form Liquid To Solid
Formula Weight 286.33
Quantity 1g
Specific Rotation Condition −4.20° (24°C c=13,absol. alcohol)
CAS Min % 96.0
CAS Max % 100.0
Specific Rotation -7.0° to -8.0° (24°C, 589nm) (c=2, DMF)
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5055 to 1.5075 (20°C, 589nm)
CAS 23788-74-1
Assay Percent Range 97%
Molecular Formula C13H18O5S
Chemical Name or Material (R)-(-)-2, 2-Dimethyl-4-(hydroxymethyl)-1, 3-dioxolane-p-toluenesulfonate
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00063233
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (R)-(-)-2, 2-Dimethyl-4-(hydroxymethyl)-1, 3-dioxolane-p-toluenesulfonate
CAS 23788-74-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H18O5S
Formula Weight 286.33
MDL Number MFCD00063233
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents