Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2,2'-Bis(N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, CTH-(R)-BINAM, 95%

Asymmetric hydrogenation

$82.50 - $330.95

Chemical Identifiers

Chemical Name or Material (R)- (+)-2,2'-Bis (N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl
CAS 208248-67-3
Name Note CTH-(R)-BINAM
Molecular Formula C44H42N2P2
Formula Weight 660.78
MDL Number MFCD04974231
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4462002

 
alfa aesar™
4462002
0.5g Each for $330.95

AA4462077

 
alfa aesar™
4462077
0.1g Each for $82.50
Description & Specifications

Specifications

Formula Weight 660.78
Chemical Name or Material (R)- (+)-2,2'-Bis (N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl
Name Note CTH-(R)-BINAM
Physical Form Powder
Assay 95%
Molecular Formula C44H42N2P2
MDL Number MFCD04974231
CAS 208248-67-3
Health Hazard 1 Warning
Quantity 0.5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under Argon

  • Packaged under Argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Packaged under Argon