Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl-3,3'-diboronic acid pinacol ester, 97%

$266.36 - $266.36

Chemical Identifiers

Chemical Name or Material (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl-3,3'-diboronic acid pinacol ester
CAS 260442-17-9
Molecular Formula C36H44B2O8
Formula Weight 626.36g/mol
MDL Number MFCD16294496
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3274503

 
alfa aesar™
H32745-03
1g Each for $266.36
Description & Specifications

Specifications

Melting Point 193° to 195°C
Quantity 1g
Chemical Name or Material (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl-3,3'-diboronic acid pinacol ester
CAS 260442-17-9
Assay 97%
Molecular Formula C36H44B2O8
Formula Weight 626.36g/mol
MDL Number MFCD16294496