Please login to your online account to display your discounted pricing

(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥95%, ACROS Organics™

$53.51 - $53.51

Chemical Identifiers

Chemical Name or Material (R)-(+)-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
CAS 99646-28-3
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 95+%
Molecular Formula C48H40P2
Formula Weight 678.79
MDL Number MFCD01311709
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382340010

  SDS
thermo fisher scientific
AC382340010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC382342500

  SDS
thermo fisher scientific
AC382342500
Glass bottle 250mg Each for $53.51
Description & Specifications

Specifications

Assay ≥95.0%
Physical Form Powder
Formula Weight 678.79
Quantity 1g
Color White to cream
Specific Rotation Condition +167.50° (20°C c=0,3%,toluene)
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 95+%
Molecular Formula C48H40P2
Specific Rotation +165° to +170° (20°C, 589nm) (C=0,3%, toluene)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (R)-(+)-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
Infrared Spectrum Authentic
MDL Number MFCD01311709
CAS 99646-28-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.