Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2,2'-Bis(di-3,5-xylylphosphino)-1,1'-binaphthyl, 98%

$468.61 - $468.61

Chemical Identifiers

Chemical Name or Material (R)-(+)-2,2'-Bis(di-3,5-xylylphosphino)-1,1'-binaphthyl
CAS 137219-86-4
Molecular Formula C52H48P2
Formula Weight 734.91
MDL Number MFCD01630821
Synonym (R)-Xylyl-BINAP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27680ME

  SDS
alfa aesar™
H27680ME
500mg Each for $468.61

AAH27680MC

  SDS
alfa aesar™
H27680MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (R)-(+)-2,2'-Bis(di-3,5-xylylphosphino)-1,1'-binaphthyl
CAS 137219-86-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C52H48P2
Formula Weight 734.91
MDL Number MFCD01630821
Synonym (R)-Xylyl-BINAP
Melting Point 201° to 203°C
Quantity 500mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.