Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine

Asymmetric hydrogenation

$81.34 - $271.99

Chemical Identifiers

Chemical Name or Material (R)- (+)-2,2',6,6'-Tetramethoxy-4,4'-bis (diphenylphosphino)-3,3'-bipyridine
CAS 221012-82-4
Molecular Formula C38H34N2O4P2
Formula Weight 644.64g/mol
MDL Number MFCD04038734
Synonym (R)-P-Phos
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4461002

 
alfa aesar™
44610-02
0.5g Each for $271.99

AA4461077

 
alfa aesar™
44610-77
0.1g Each for $81.34
Description & Specifications

Specifications

Quantity 0.5g
Melting Point 261° to 265°C
Quantity 0.5g
Odor Odorless
Chemical Name or Material (R)- (+)-2,2',6,6'-Tetramethoxy-4,4'-bis (diphenylphosphino)-3,3'-bipyridine
CAS 221012-82-4
Physical Form Powder
Molecular Formula C38H34N2O4P2
Formula Weight 644.64g/mol
MDL Number MFCD04038734
Synonym (R)-P-Phos
Storage Note 1
  • Packaged under Argon

  • Packaged under Argon

Safety and Handling

  • Packaged under Argon