Please login to your online account to display your discounted pricing

R 1530, Tocris™

Multi-RTK inhibitor; inhibits angiogenesis

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material R 1530
Assay Percent Range >98%
Molecular Formula C18H14ClFN4O
Formula Weight 356.78
Synonym 5-(2-Chlorophenyl)-7-fluoro-1,2-dihydro-8-methoxy-3-methylpyrazolo[3,4-b][1,4]benzodiazepine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

488510

 
r & d systems
488510
10mg Each for $257.85

488550

 
r & d systems
488550
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 356.78
Quantity 10mg
Chemical Name or Material R 1530
Assay Percent Range >98%
Molecular Formula C18H14ClFN4O
CAS 882531-87-5
Synonym 5-(2-Chlorophenyl)-7-fluoro-1,2-dihydro-8-methoxy-3-methylpyrazolo[3,4-b][1,4]benzodiazepine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C