Please login to your online account to display your discounted pricing

(R)-(-)-10-Methyl-1(9)-octal-2-one, 98.5%, ACROS Organics™


Quantity 250mg
Packaging Glass Bottle in Plastic Container

Manufacturer: thermo fisher scientific  240812500

 View more versions of this product

Catalog No. AC240812500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Liquid
Formula Weight 164.25
Quantity 250mg
Color Clear colorless to yellow
Packaging Glass Bottle in Plastic Container
TSCA TSCA
Density 1.01g/mL
Specific Rotation Condition −207° (20°C C=1,ethanol)
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material (R)-(-)-10-Methyl-1(9)-octal-2-one
Infrared Spectrum Authentic
Refractive Index 1.5235 to 1.5255 (20°C, 589nm)
Specific Rotation -202° to -212° (20°C, 589nm) (c=1, ethanol)
CAS 63975-59-7
Synonym (R)-(-)-4,4a,5,6,7,8-Hexahydro-4a-methyl-2(3H)-naphthalenone
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98.5%
Assay Percent Range 98.50%
Molecular Formula C11H16O
MDL Number MFCD00009959

Specifications

Chemical Name or Material (R)-(-)-10-Methyl-1(9)-octal-2-one
Name Note 98.5%
CAS 63975-59-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98.50%
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00009959
Synonym (R)-(-)-4,4a,5,6,7,8-Hexahydro-4a-methyl-2(3H)-naphthalenone
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents