Please login to your online account to display your discounted pricing

(R)-(-)-10-Methyl-1(9)-octal-2-one, 98.5%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-(-)-10-Methyl-1(9)-octal-2-one
Name Note 98.5%
CAS 63975-59-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98.50%
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00009959
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC240812500

  SDS
thermo fisher scientific
240812500
Glass Bottle in Plastic Container 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Liquid
Formula Weight 164.25
Quantity 250mg
Color Clear colorless to yellow
Packaging Glass Bottle in Plastic Container
TSCA TSCA
Density 1.01g/mL
Specific Rotation Condition −207° (20°C C=1,ethanol)
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material (R)-(-)-10-Methyl-1(9)-octal-2-one
Infrared Spectrum Authentic
Refractive Index 1.5235 to 1.5255 (20°C, 589nm)
Specific Rotation -202° to -212° (20°C, 589nm) (c=1, ethanol)
CAS 63975-59-7
Synonym (R)-(-)-4,4a,5,6,7,8-Hexahydro-4a-methyl-2(3H)-naphthalenone
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98.5%
Assay Percent Range 98.50%
Molecular Formula C11H16O
MDL Number MFCD00009959
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.