Please login to your online account to display your discounted pricing

(R)-(+)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-(+)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone
Name Note 99%
CAS 119838-44-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13 H24 N2 O3
Formula Weight 256.35
MDL Number MFCD00075100
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC320562500

  SDS
thermo fisher scientific
320562500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 256.35
Quantity 250mg
Color White
TSCA TSCA
Melting Point 66° to 70°C
CAS Max % 100.0
Name Note 99%
Assay Percent Range 99%
Molecular Formula C13 H24 N2 O3
Specific Rotation +13° to +16° (20°C, 589nm) (c=1, CH2Cl2)
CAS Min % 98.5
Chemical Name or Material (R)-(+)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone
Packaging Glass bottle
MDL Number MFCD00075100
CAS 119838-44-7
Infrared Spectrum Authentic