Please login to your online account to display your discounted pricing

(R)-(+)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone, 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  320562500

 View more versions of this product

Catalog No. AC320562500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 256.35
Quantity 250mg
Color White
TSCA TSCA
Melting Point 66° to 70°C
CAS Max % 100.0
Name Note 99%
Assay Percent Range 99%
Molecular Formula C13 H24 N2 O3
Specific Rotation +13° to +16° (20°C, 589nm) (c=1, CH2Cl2)
CAS Min % 98.5
Chemical Name or Material (R)-(+)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone
Packaging Glass bottle
MDL Number MFCD00075100
CAS 119838-44-7
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (R)-(+)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone
Name Note 99%
CAS 119838-44-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13 H24 N2 O3
Formula Weight 256.35
MDL Number MFCD00075100