Please login to your online account to display your discounted pricing

(R)-(-)-1-Methyl-3-phenylpropylamine, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC414760050

 View more versions of this product

Catalog No. AC414760050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material (R)-(-)-1-Methyl-3-phenylpropylamine
Density 0.9360g/mL
Specific Rotation Condition − 11.00 (20.00°C neat)
Color Clear colorless
Quantity 5g
CAS 937-52-0
Infrared Spectrum Authentic
Refractive Index 1.5130 to 1.5150 (20°C, 589nm)
Specific Rotation −10° to −12° (20°C, 589nm) (neat)
Physical Form Liquid
Assay ≥97.5% (total base)
Assay Percent Range 98%
Molecular Formula C10H15N
Formula Weight 149.24

Specifications

Chemical Name or Material (R)-(-)-1-Methyl-3-phenylpropylamine
Assay ≥97.5% (total base)
Assay Percent Range 98%
Molecular Formula C10H15N
Formula Weight 149.24