missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-Fmoc-4-oxopiperidine-2-carboxylic acid, 97%

$91.30 - $279.12

Chemical Identifiers

CAS 1375078-38-8
Molecular Formula C21H19NO5
Molecular Weight (g/mol) 365.385
MDL Number MFCD22208545
InChI Key YGAHCGORXONSSM-LJQANCHMSA-N
Synonym r-1-fmoc-4-oxopiperidine-2-carboxylic acid, 2r-1-9h-fluoren-9-ylmethoxy carbonyl-4-oxopiperidine-2-carboxylic acid
PubChem CID 92174274
IUPAC Name (2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
SMILES C1CN(C(CC1=O)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66328MD View Documents Alfa Aesar™
H66328MD
250mg Each for $91.30
 
AAH6632803 View Documents Alfa Aesar™
H6632803
1g Each for $279.12
 
Specifications

Chemical Identifiers

1375078-38-8
365.385
YGAHCGORXONSSM-LJQANCHMSA-N
92174274
C1CN(C(CC1=O)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C21H19NO5
MFCD22208545
r-1-fmoc-4-oxopiperidine-2-carboxylic acid, 2r-1-9h-fluoren-9-ylmethoxy carbonyl-4-oxopiperidine-2-carboxylic acid
(2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-oxopiperidine-2-carboxylic acid

Specifications

Moisture Sensitive
250mg
C21H19NO5
r-1-fmoc-4-oxopiperidine-2-carboxylic acid, 2r-1-9h-fluoren-9-ylmethoxy carbonyl-4-oxopiperidine-2-carboxylic acid
C1CN(C(CC1=O)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
365.385
365.38
97%
White to Yellow
1375078-38-8
MFCD22208545
YGAHCGORXONSSM-LJQANCHMSA-N
(2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
92174274
Powder
(R)-(+)-1-Fmoc-4-oxopiperidine-2-carboxylic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C