Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%

Chemical Identifiers

Chemical Name or Material (R)-1-Cyclopropylethylamine
CAS 6240-96-6
Molecular Formula C5H11N
Formula Weight 85.15
MDL Number MFCD08064288
UN Number UN2733
Synonym (R)-alpha-Methylcyclopropanemethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2690203

 
alfa aesar™
H2690203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2690206

 
alfa aesar™
H2690206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 85.15
Chemical Name or Material (R)-1-Cyclopropylethylamine
Assay 98%
Assay Percent Notes ee ≥98%
Molecular Formula C5H11N
MDL Number MFCD08064288
CAS 6240-96-6
UN Number UN2733
Synonym (R)-alpha-Methylcyclopropanemethylamine
Melting Point −68°C
pH 13
Health Hazard 1 Danger
Boiling Point 95°C
Quantity 1g
Flash Point 3°C (37°F)
Odor Amine-like
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store in a well-ventilated place.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

Applications
(R)-1-Cyclopropylethylamine is used for resolution of organic compounds in organic synthesis.

Solubility
Miscible with water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store in a well-ventilated place.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.