Learn More
(R)-(-)-1-Cyclohexylethylamine, ChiPros 98%, ee 94+%, Thermo Scientific Chemicals
$51.59 - $361.38
Chemical Identifiers
CAS | 5913-13-3 |
---|---|
Molecular Formula | C8H17N |
Molecular Weight (g/mol) | 127.23 |
MDL Number | MFCD00043338 |
InChI Key | XBWOPGDJMAJJDG-UHFFFAOYNA-N |
Synonym | r---1-cyclohexylethylamine, r-1-cyclohexylethylamine, 1r-1-cyclohexylethanamine, r-1-cyclohexylethanamine, 1r-1-cyclohexylethylamine, chembl19290, 1r-1-cyclohexylethan-1-amine, 1-cyclohexyl-ethylamine, 1-cyclohexylethanamine #, r---cyclohexylethylamine |
PubChem CID | 10997046 |
SMILES | CC(N)C1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1905103
|
Thermo Scientific Chemicals
L1905103 |
1 g |
Each for $51.59
|
|
|||||
AAL1905106
|
Thermo Scientific Chemicals
L1905106 |
5 g |
Each for $102.96
|
|
|||||
AAL1905114
|
Thermo Scientific Chemicals
L1905114 |
25 g |
Each for $361.38
|
|
|||||
Description
(R)-(-)-1-Cyclohexylethylamine is used in the preparation of benzylidene(1-cyclohexylethyl)amine by reacting with benzaldehyde. It is also used to prepare chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides. In addition to this, it is used in the preparation of polyamides bound to silica gel for high performance liquid chromatography, which are pressure-stable.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(-)-1-Cyclohexylethylamine is used in the preparation of benzylidene(1-cyclohexylethyl)amine by reacting with benzaldehyde. It is also used to prepare chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides. In addition to this, it is used in the preparation of polyamides bound to silica gel for high performance liquid chromatography, which are pressure-stable.
Solubility
Miscible with acetone, chloroform,dimethyl sulfoxide, ethanol, methanol, ether, petroleum ether and dichloromethane. Insoluble in water.
Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
5913-13-3 | |
127.23 | |
XBWOPGDJMAJJDG-UHFFFAOYNA-N | |
10997046 |
C8H17N | |
MFCD00043338 | |
r---1-cyclohexylethylamine, r-1-cyclohexylethylamine, 1r-1-cyclohexylethanamine, r-1-cyclohexylethanamine, 1r-1-cyclohexylethylamine, chembl19290, 1r-1-cyclohexylethan-1-amine, 1-cyclohexyl-ethylamine, 1-cyclohexylethanamine #, r---cyclohexylethylamine | |
CC(N)C1CCCCC1 |
Specifications
5913-13-3 | |
11 | |
51°C (123°F) | |
C8H17N | |
MFCD00043338 | |
UN2733 | |
Air Sensitive | |
Miscible with acetone,chloroform,dimethyl sulfoxide,ethanol,methanol,ether,petroleum ether and dichloromethane. Insoluble in water. | |
−4° (Neat) | |
(1R)-1-cyclohexylethanamine | |
10997046 | |
98% | |
ee ≥94% |
0.866 | |
177°C to 178°C | |
Amine-like | |
1.462 | |
1 g | |
2935070 | |
r---1-cyclohexylethylamine, r-1-cyclohexylethylamine, 1r-1-cyclohexylethanamine, r-1-cyclohexylethanamine, 1r-1-cyclohexylethylamine, chembl19290, 1r-1-cyclohexylethan-1-amine, 1-cyclohexyl-ethylamine, 1-cyclohexylethanamine #, r---cyclohexylethylamine | |
XBWOPGDJMAJJDG-UHFFFAOYNA-N | |
CC(N)C1CCCCC1 | |
127.23 | |
127.23 | |
(R)-(-)-1-Cyclohexylethylamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 227-634-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only