Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylic acid, 95%

$237.14 - $237.14

Chemical Identifiers

Chemical Name or Material (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylic acid
CAS 1212176-33-4
Molecular Formula C11H17NO5
Formula Weight 243.26g/mol
MDL Number MFCD09953681
Synonym (R)-1-Boc-4-oxopipecolic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25903ME

 
alfa aesar™
H25903-ME
500mg Each for $237.14
Description & Specifications

Specifications

Quantity 500mg
Melting Point ≈115°C (dec.)
Quantity 500mg
Optical Rotation +22° (c=1 in Chloroform)
Chemical Name or Material (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylic acid
CAS 1212176-33-4
Assay 95%
Molecular Formula C11H17NO5
Formula Weight 243.26g/mol
MDL Number MFCD09953681
Synonym (R)-1-Boc-4-oxopipecolic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.