missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-1-BOC-3-methylpiperazine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC432050050

 View more versions of this product

Catalog No. AC432050050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder
≥98.5%
White
100.0
Authentic
C10H20N2O2
MFCD02683205
98.5
Glass bottle
200.28
5g
43° to 47°C
tert-Butyl (R)-3-methyl-1-piperazinecarboxylate
(R)-1-BOC-3-Methylpiperazine
163765-44-4
99%
+14° to +18° (20°C, 589nm) (c=1, chloroform)

Chemical Identifiers

163765-44-4
100.0
C10H20N2O2
MFCD02683205
(R)-1-BOC-3-Methylpiperazine
98.5
99%
200.28
tert-Butyl (R)-3-methyl-1-piperazinecarboxylate
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.