missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-1-BOC-3-hydroxypiperidine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436430010
Specifications
(R)-1-BOC-3-Hydroxypiperidine | |
Authentic | |
-8.5° to -12.0° (20°C, 589nm) (c=1 in CHCl3) | |
40.0°C to 42.0°C | |
143900-43-0 | |
100.0 | |
C10H19NO3 | |
UIJXHKXIOCDSEB-MRVPVSSYSA-N | |
tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate | |
1514398 | |
Low Melting Solid |
104.4°C | |
Glass bottle | |
Tan to White | |
1g | |
96.0 | |
96% min. (GC) | |
1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, r-1-boc-3-hydroxypiperidine, r-1-n-boc-3-hydroxypiperidine, r-3-hydroxy-1-tert-butoxycarbonyl piperidine, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-n-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate | |
CC(C)(C)OC(=O)N1CCCC(C1)O | |
201.26 | |
201.26 | |
97% |
Chemical Identifiers
143900-43-0 | |
201.26 | |
1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, r-1-boc-3-hydroxypiperidine, r-1-n-boc-3-hydroxypiperidine, r-3-hydroxy-1-tert-butoxycarbonyl piperidine, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-n-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate | |
tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate |
C10H19NO3 | |
UIJXHKXIOCDSEB-MRVPVSSYSA-N | |
1514398 | |
CC(C)(C)OC(=O)N1CCCC(C1)O |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger