missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-1-BOC-3-(hydroxymethyl)piperidine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436480010
Specifications
(R)-1-BOC-3-(Hydroxymethyl)piperidine | |
Glass bottle | |
White to Yellow | |
1g | |
96.0 | |
96% min. (GC) | |
r---1-boc-3-hydroxymethyl piperidine, r-1-boc-3-hydroxymethyl piperidine, r-1-boc-3-hyroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl-piperidine, r-n-boc-3-hydroxymethyl piperidine, r-n-boc-3-piperidinemethanol, r-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, tert-butyl 3r-3-hydroxymethyl piperidine-1-carboxylate | |
CC(C)(C)OC(=O)N1CCCC(C1)CO | |
215.29 | |
215.29 | |
97% |
Authentic | |
-11.2 | |
92°C to 93°C | |
140695-85-8 | |
100.0 | |
C11H21NO3 | |
OJCLHERKFHHUTB-SECBINFHSA-N | |
tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate | |
1514447 | |
Powder |
Chemical Identifiers
140695-85-8 | |
215.29 | |
r---1-boc-3-hydroxymethyl piperidine, r-1-boc-3-hydroxymethyl piperidine, r-1-boc-3-hyroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl-piperidine, r-n-boc-3-hydroxymethyl piperidine, r-n-boc-3-piperidinemethanol, r-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, tert-butyl 3r-3-hydroxymethyl piperidine-1-carboxylate | |
tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate |
C11H21NO3 | |
OJCLHERKFHHUTB-SECBINFHSA-N | |
1514447 | |
CC(C)(C)OC(=O)N1CCCC(C1)CO |