Please login to your online account to display your discounted pricing

(R)-1-BOC-3-(hydroxymethyl)piperidine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-1-BOC-3-(Hydroxymethyl)piperidine
CAS 140695-85-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H21NO3
Formula Weight 215.29
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC436480010

  SDS
thermo fisher scientific
AC436480010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC436480050

  SDS
thermo fisher scientific
AC436480050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 215.29
Quantity 1g
Color White to yellow
Melting Point 90° to 95°C
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C11H21NO3
CAS 140695-85-8
Chemical Name or Material (R)-1-BOC-3-(Hydroxymethyl)piperidine
Specific Rotation -8° to -12° (20°C, 589nm) (c=1, chloroform)
Infrared Spectrum Authentic