Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-1-Boc-3-fluoropyrrolidine, 96%

Chemical Identifiers

Chemical Name or Material (R)-(-)-1-Boc-3-fluoropyrrolidine
CAS 876617-25-3
Formula Weight 189.23
MDL Number MFCD04114065
UN Number UN2810
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6653403

  SDS
alfa aesar™
H6653403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66534MD

  SDS
alfa aesar™
H66534MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 85° to 95°C (2mmHg)
Formula Weight 189.23
Chemical Name or Material (R)-(-)-1-Boc-3-fluoropyrrolidine
MDL Number MFCD04114065
Physical Form Liquid
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Sensitivity Moisture Sensitive
CAS 876617-25-3
Health Hazard 1 Danger
Color Colorless to pale yellow
Health Hazard 2 GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Refractive Index 1.4420
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
UN Number UN2810
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.