Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine, 97%

Chemical Identifiers

Chemical Name or Material (R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine
CAS 199174-29-3
Formula Weight 200.28
MDL Number MFCD03844734
Beilstein 10537871
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6627703

 
alfa aesar™
H6627703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66277MD

 
alfa aesar™
H66277MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.28
CAS 199174-29-3
Chemical Name or Material (R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine
Physical Form Liquid
MDL Number MFCD03844734
Assay 97%
Refractive Index 1.4780
Quantity 1g
Color Colorless to yellow
Beilstein 10537871
Storage Note 1 2° to 8°C
Sensitivity Air Sensitive
Safety and Handling

2° to 8°C