missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-1-BOC-3-(Aminomethyl)piperidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436420050

 View more versions of this product

Catalog No. AC436420050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(R)-1-BOC-3-(Aminomethyl)piperidine
Glass bottle
5g
96.0
96% min. (GC)
3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine, r-1-boc-3-aminomethyl piperidine, r-1-n-boc-3-aminomethyl piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)CN
214.31
214.31
97%
Authentic
Colorless to Yellow
140645-23-4
100.0
C11H22N2O2
WPWXYQIMXTUMJB-SECBINFHSA-N
tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate
1502023
Liquid

Chemical Identifiers

140645-23-4
214.31
3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine, r-1-boc-3-aminomethyl piperidine, r-1-n-boc-3-aminomethyl piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate
tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate
C11H22N2O2
WPWXYQIMXTUMJB-SECBINFHSA-N
1502023
CC(C)(C)OC(=O)N1CCCC(C1)CN
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

SDS
Documents