Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-1-Benzyloxycarbonyl-5-pyrrolidinone-2-carboxylic acid, 95%

Chemical Identifiers

Chemical Name or Material (R)-1-Benzyloxycarbonyl-5-pyrrolidinone-2-carboxylic acid
CAS 78339-57-8
Formula Weight 263.25
MDL Number MFCD00077036
Beilstein 4698210
Synonym Z-D-Pyr-OH; N-Benzyloxycarbonyl-D-pyroglutamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6641706

 
alfa aesar™
H6641706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6641714

 
alfa aesar™
H6641714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 138° to 144°C
Formula Weight 263.25
CAS 78339-57-8
Physical Form Powder
MDL Number MFCD00077036
Beilstein 4698210
Synonym Z-D-Pyr-OH; N-Benzyloxycarbonyl-D-pyroglutamic acid
Assay 95%
Color White
Storage Note 1 -30° to -10°C
Chemical Name or Material (R)-1-Benzyloxycarbonyl-5-pyrrolidinone-2-carboxylic acid
Quantity 5g
Safety and Handling

-30° to -10°C