missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(-)-1-Benzyl-3-aminopyrrolidine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437890050
Specifications
(R)-(-)-1-Benzyl-3-aminopyrrolidine | |
140°C | |
1.02 | |
108°C to 115°C (5.0mmHg) | |
5g | |
98.5 | |
98.5% min. (GC) | |
3-pyrrolidinamine, 1-phenylmethyl-, 3r, 3r---1-benzyl-3-aminopyrrolidine, 3r-1-benzyl-3-pyrrolidinamine, 3r-1-benzylpyrrolidin-3-amine, r---1-benzyl-3-aminopyrrolidine, r-1-benzyl-3-aminopyrrolidine, r-1-benzylpyrrolidin-3-amine, r-3-amino-1-benzylpyrrolidine, r-3-amino-1-n-benzyl-pyrrolidine, r-n-benzyl-3-aminopyrrolidine | |
C1CN(CC1N)CC2=CC=CC=C2 | |
176.26 | |
176.26 | |
99% |
1.0200g/mL | |
Authentic | |
-2.2 | |
Colorless to Yellow | |
114715-39-8 | |
100.0 | |
C11H16N2 | |
HBVNLKQGRZPGRP-LLVKDONJSA-N | |
(3R)-1-benzylpyrrolidin-3-amine | |
1519354 | |
Liquid |
Chemical Identifiers
114715-39-8 | |
176.26 | |
3-pyrrolidinamine, 1-phenylmethyl-, 3r, 3r---1-benzyl-3-aminopyrrolidine, 3r-1-benzyl-3-pyrrolidinamine, 3r-1-benzylpyrrolidin-3-amine, r---1-benzyl-3-aminopyrrolidine, r-1-benzyl-3-aminopyrrolidine, r-1-benzylpyrrolidin-3-amine, r-3-amino-1-benzylpyrrolidine, r-3-amino-1-n-benzyl-pyrrolidine, r-n-benzyl-3-aminopyrrolidine | |
(3R)-1-benzylpyrrolidin-3-amine |
C11H16N2 | |
HBVNLKQGRZPGRP-LLVKDONJSA-N | |
1519354 | |
C1CN(CC1N)CC2=CC=CC=C2 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger