Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-1-Benzoyl-3-methylpiperazine hydrochloride, 99%, ee 99%

$95.43 - $1,059.03

Chemical Identifiers

Chemical Name or Material (R)-1-Benzoyl-3-methylpiperazine hydrochloride
CAS 474010-60-1
Molecular Formula C12H16N2O·HCl
Formula Weight 240.73
MDL Number MFCD05664046
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2005903

  SDS
alfa aesar™
L2005903
1g Each for $270.92

AAL2005906

  SDS
alfa aesar™
L2005906
5g Each for $1,059.03

AAL20059MD

  SDS
alfa aesar™
L20059MD
250mg Each for $95.43
Description & Specifications

Specifications

Formula Weight 240.73
Chemical Name or Material (R)-1-Benzoyl-3-methylpiperazine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Assay Percent Notes ee 99%
Molecular Formula C12H16N2O·HCl
MDL Number MFCD05664046
CAS 474010-60-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
(R)-1-Benzoyl-3-methylpiperazine hydrochloride is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.