Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-1-Aminoindane, ChiPros 99+%, ee 98+%

$120.20 - $443.31

Chemical Identifiers

Chemical Name or Material (R)-(-)-1-Aminoindane
CAS 10277-74-4
Molecular Formula C9H11N
Formula Weight 133.19
MDL Number MFCD00216669
Beilstein 3196204
Synonym (R)-(-)-1-Indanamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1930203

  SDS
alfa aesar™
L1930203
1g Each for $120.20

AAL1930206

  SDS
alfa aesar™
L1930206
5g Each for $443.31
Description & Specifications

Specifications

Formula Weight 133.19
Chemical Name or Material (R)-(-)-1-Aminoindane
Assay ≥99%
CAS 10277-74-4
Assay Percent Notes ee ≥98%
Molecular Formula C9H11N
MDL Number MFCD00216669
Beilstein 3196204
Synonym (R)-(-)-1-Indanamine
Health Hazard 1 Warning
Boiling Point 225°C
Quantity 1g
Flash Point 94°C (201°F)
Odor Amine-like
Optical Rotation −16.5° (c=1.5 in Methanol)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
(R)-(-)-1-Aminoindane is used as an intermediate of N-[1-(R)-indanyl]adenosine. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

Solubility
Soluble in methanol. Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to air. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.