Please login to your online account to display your discounted pricing

(R)-(-)-1-Aminoindan, 97%, ACROS Organics™

$349.45 - $349.45

Chemical Identifiers

Chemical Name or Material (R)-(-)-1-Aminoindan
CAS 10277-74-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H11N
Formula Weight 133.19
MDL Number MFCD00216669
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC342520010

  SDS
thermo fisher scientific
342520010
Glass bottle 1g Each for $349.45
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 133.19
Physical Form Liquid
Quantity 1g
Color Clear colorless to slightly yellow
CAS Max % 100.0
CAS Min % 96.0
Refractive Index 1.5600 to 1.5620 (20°C, 589nm)
Chemical Name or Material (R)-(-)-1-Aminoindan
Specific Rotation -15° to -25° (20°C, 589nm) (c=1.5, CH3OH)
Molecular Formula C9H11N
CAS 10277-74-4
Assay Percent Range 97%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00216669
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.