Please login to your online account to display your discounted pricing

(R)-(-)-1-Amino-2-propanol, +98%, ACROS Organics™

$58.14 - $258.72

Chemical Identifiers

Chemical Name or Material (R)-(-)-1-Amino-2-propanol
CAS 2799-16-8
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C3H9NO
Linear Formula CH3CH(OH)CH2NH2
Formula Weight 75.11
MDL Number MFCD00064428
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC218970250

  SDS
thermo fisher scientific
AC218970250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC218970050

  SDS
thermo fisher scientific
218970050
Glass bottle 5g Each for $258.72

AC218970010

  SDS
thermo fisher scientific
218970010
Glass bottle 1g Each for $58.14
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 75.11
Physical Form Liquid After Melting
Quantity 25g
Color Clear colorless to light yellow
Density 0.9540g/mL
Specific Rotation Condition − 18.00 (20.00°C c=1.8,H2O)
Beilstein 04, II, 736
CAS Max % 100.0
CAS Min % 98.0
Synonym (R)-(-)-Isopropanolamine
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00064428
Chemical Name or Material (R)-(-)-1-Amino-2-propanol
Molecular Formula C3H9NO
CAS 2799-16-8
Assay Percent Range 98+%
Specific Rotation -16° to -20° (20°C, 589nm) (c=1.8, H2O)
Infrared Spectrum Authentic
Refractive Index 1.4465 to 1.4495 (20°C, 589nm)
Linear Formula CH3CH(OH)CH2NH2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.