Please login to your online account to display your discounted pricing

(R)-(-)-1-Amino-2-propanol, +98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  218970050

 View more versions of this product

Catalog No. AC218970050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 75.11
Physical Form Liquid After Melting
Quantity 5g
Color Clear colorless to light yellow
Beilstein 04, II, 736
CAS Min % 98.0
CAS Max % 100.0
Synonym (R)-(-)-Isopropanolamine
Linear Formula CH3CH(OH)CH2NH2
Specific Rotation -16° to -20° (20°C, 589nm) (c=1.8, H2O)
Refractive Index 1.4465 to 1.4495 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00064428
Assay Percent Range 98+%
Chemical Name or Material (R)-(-)-1-Amino-2-propanol
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Molecular Formula C3H9NO
CAS 2799-16-8
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (R)-(-)-1-Amino-2-propanol
CAS 2799-16-8
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C3H9NO
Linear Formula CH3CH(OH)CH2NH2
Formula Weight 75.11
MDL Number MFCD00064428
Synonym (R)-(-)-Isopropanolamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents