Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-(4-Methylphenyl)ethylamine, ChiPros 98+%, ee 98%

Chemical Identifiers

Chemical Name or Material (R)-(+)-1-(4-Methylphenyl)ethylamine
CAS 4187-38-6
Molecular Formula C9H13N
Formula Weight 135.21g/mol
MDL Number MFCD00145202
UN Number UN2735
Beilstein 3195425
Synonym (R)-alpha,p-Dimethylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1632803

 
alfa aesar™
L16328-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1632806

 
alfa aesar™
L16328-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1632814

 
alfa aesar™
L16328-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Boiling Point 204°C
Melting Point <−20°C
Quantity 1g
Flash Point 82°C (179°F)
Odor Amine-like
Chemical Name or Material (R)-(+)-1-(4-Methylphenyl)ethylamine
CAS 4187-38-6
Assay ≥98%
Assay Percent Notes ee 98%
Molecular Formula C9H13N
Formula Weight 135.21g/mol
MDL Number MFCD00145202
UN Number UN2735
Beilstein 3195425
Synonym (R)-alpha,p-Dimethylbenzylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.