missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-1-(4-Methoxyphenyl)ethylamine, TCI America™

$76.39 - $232.23

Chemical Identifiers

CAS 22038-86-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671659
InChI Key JTDGKQNNPKXKII-SSDOTTSWSA-N
Synonym r-+-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethanamine, 1r-1-4-methoxyphenyl ethanamine, r-+-4-methoxy-alpha-methylbenzylamine, s-4-methoxy-alpha-methylbenzylamine, r +-1-4-methylphenyl ethylamine, pubchem9920, ksc496q8b, rm+-1-4-methoxyphenyl ethylamine
PubChem CID 641449
IUPAC Name (1R)-1-(4-methoxyphenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)OC)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M15115G View Documents TCI America
M15115G
5g Each for $76.39
 
M151125G View Documents TCI America
M151125G
25g Each for $232.23
 
Specifications

Chemical Identifiers

22038-86-4
151.209
JTDGKQNNPKXKII-SSDOTTSWSA-N
641449
CC(C1=CC=C(C=C1)OC)N
C9H13NO
MFCD00671659
r-+-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethanamine, 1r-1-4-methoxyphenyl ethanamine, r-+-4-methoxy-alpha-methylbenzylamine, s-4-methoxy-alpha-methylbenzylamine, r +-1-4-methylphenyl ethylamine, pubchem9920, ksc496q8b, rm+-1-4-methoxyphenyl ethylamine
(1R)-1-(4-methoxyphenyl)ethanamine

Specifications

Yellow
22038-86-4
MFCD00671659
r-+-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethanamine, 1r-1-4-methoxyphenyl ethanamine, r-+-4-methoxy-alpha-methylbenzylamine, s-4-methoxy-alpha-methylbenzylamine, r +-1-4-methylphenyl ethylamine, pubchem9920, ksc496q8b, rm+-1-4-methoxyphenyl ethylamine
CC(C1=CC=C(C=C1)OC)N
151.209
151.21
≥98.0% (GC,T)
5g
C9H13NO
2735
JTDGKQNNPKXKII-SSDOTTSWSA-N
(1R)-1-(4-methoxyphenyl)ethanamine
641449
Clear Liquid at 20°C
(R)-(+)-1-(4-Methoxyphenyl)ethylamine
Safety and Handling

Safety and Handling

TSCA : No