Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-(4-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%


Quantity 5g

Manufacturer: alfa aesar™  L1632106

 View more versions of this product

Catalog No. AAL1632106

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 151.21
Chemical Name or Material (R)-(+)-1-(4-Methoxyphenyl)ethylamine
Synonym (R)-p-Methoxy-alpha-methylbenzylamine
Assay ≥99%
Beilstein 2413029
Health Hazard 2 GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
pH 11
Odor Amine-like
Optical Rotation +34° (Neat)
MDL Number MFCD00671659
CAS 22038-86-4
Health Hazard 1 Danger
Molecular Formula C9H13NO
Boiling Point 65°C (0.38mmHg)
Melting Point <−20°C
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay Percent Notes ee ≥99%
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
UN Number UN2735
Quantity 5g

Specifications

Chemical Name or Material (R)-(+)-1-(4-Methoxyphenyl)ethylamine
CAS 22038-86-4
Molecular Formula C9H13NO
Formula Weight 151.21
MDL Number MFCD00671659
UN Number UN2735
Beilstein 2413029
Synonym (R)-p-Methoxy-alpha-methylbenzylamine

Applications
(R)-(+)-1-(4-Methoxyphenyl)ethylamine use in diastereo-and enantioselective Michael addition reactions. Thiazoles has been prepared from (R)-(+)-1-(4-Methoxyphenyl)ethylamine.

Solubility
Soluble in water (10g/L).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.

Accessories

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents