Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 98%

Chemical Identifiers

Chemical Name or Material (R)-(+)-1-(4-Fluorophenyl)ethylamine
CAS 374898-01-8
Molecular Formula C8H10FN
Formula Weight 139.17
MDL Number MFCD03093090
UN Number UN2735
Beilstein 8614058
Synonym (R)-4-Fluoro-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1912003

  SDS
alfa aesar™
L1912003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1912006

  SDS
alfa aesar™
L1912006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1912014

  SDS
alfa aesar™
L1912014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.17
CAS 374898-01-8
Assay 99%
Molecular Formula C8H10FN
Chemical Name or Material (R)-(+)-1-(4-Fluorophenyl)ethylamine
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Assay Percent Notes ee 98%
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 68°C (154°F)
Synonym (R)-4-Fluoro-alpha-methylbenzylamine
Boiling Point 76°C (22mmHg)
Melting Point −30°C
Quantity 1g
MDL Number MFCD03093090
Health Hazard 2 GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.
Beilstein 8614058
Optical Rotation +33° (Neat)
UN Number UN2735

Applications
(R)-(+)-1-(4-Fluorophenyl)ethylamine, is used as a pharmaceutical intermediate and as a chemical in organic synthesis

Solubility
Sparingly Soluble in water (1.5E-3 g/L) (25°C).

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.