Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-1-(3,4-Dimethoxyphenyl)ethylamine, ChiPros™, 98%, ee 98+%

Chemical Identifiers

Chemical Name or Material (R)-1-(3,4-Dimethoxyphenyl)ethylamine
CAS 100570-24-9
Molecular Formula C10H15NO2
Formula Weight 181.24
MDL Number MFCD06761897
UN Number UN2735
Synonym (R)-3,4-Dimethoxy-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2748403

  SDS
alfa aesar™
H2748403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.24
Boiling Point 120°C (1mmHg)
Synonym (R)-3,4-Dimethoxy-alpha-methylbenzylamine
Health Hazard 1 Danger
Quantity 1g
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (R)-1-(3,4-Dimethoxyphenyl)ethylamine
Assay Percent Notes ee ≥98%
Molecular Formula C10H15NO2
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
MDL Number MFCD06761897
CAS 100570-24-9
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 98%
UN Number UN2735
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.