Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-(2-Naphthyl)ethylamine, ChiPros 99+%, ee 99+%

Chemical Identifiers

Chemical Name or Material (R)-(+)-1-(2-Naphthyl)ethylamine
CAS 3906-16-9
Molecular Formula C12H13N
Formula Weight 171.24
MDL Number MFCD00085367
UN Number UN3259
Beilstein 3195950
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1702303

  SDS
alfa aesar™
L1702303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.24
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00085367
Chemical Name or Material (R)-(+)-1-(2-Naphthyl)ethylamine
Beilstein 3195950
UN Number UN3259
Optical Rotation +20° (c=2 in Methanol)
Boiling Point 90°C (0.15mmHg)
Quantity 1g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Assay ≥99%
Melting Point 45° to 52°C
Molecular Formula C12H13N
Health Hazard 1 Danger
Assay Percent Notes ee ≥99%
CAS 3906-16-9

Applications
(R)-(+)-1-(2-Naphthyl)ethylamine is used in studies about aldol reactions of the acetamide and reactions with the corresponding amidocuprate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air. Incompatible with acids and oxidizing agents.